Merck
  • Rapid synthesis of [18F]fluoroestradiol: remarkable advantage of microwaving over conventional heating.
Rapid synthesis of [18F]fluoroestradiol: remarkable advantage of microwaving over conventional heating.
Journal of labelled compounds & radiopharmaceuticals (2014-12-06)
Jianfeng Shi, George Afari, Sibaprasad Bhattacharyya
ABSTRACT
16α-[(18)F]fluoroestradiol ([(18)F]FES) is known as a clinically important tracer in nuclear medicine as an estrogen receptor ligand for investigating primary and metastatic breast cancers. Synthesizing [(18)F]FES is a two-step process associated with [(18)F]fluoride incorporation to the precursor (3-methoxymethyl 16β,17β-epiestriol-O-cyclic sulfone) and subsequent hydrolysis of the [(18)F]fluorinated intermediate with 2 N HCl. The impact of microwave (MW) heating on both fluorination and hydrolysis reactions was investigated. The duration and temperatures of the fluorination reaction were varied for both MW heating and conventional heating (CH) methods. Chemical and radiochemical purity and radiochemical yields were investigated for CH and compared with MW-assisted radiosyntheses. Quality control tests of MW-assisted [(18)F]FES were performed following US Pharmacopeia procedures for clinical-grade positron emission tomography pharmaceuticals. The results demonstrate that microwaving not only improves the (18)F-fluoride incorporation (~55% improvement at 110°C for 4 min) but also significantly reduces hydrolysis time (approximately sevenfold reduction at 120°C) in comparison with CH under similar conditions. The overall isolated radiochemical yield of purified [(18)F]FES was significantly higher (~90% improvement) with MW, and side products were notably fewer. Quality control test results demonstrated that [(18)F]FES produced by microwaving was suitable for human injection.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture