Merck
  • HPLC-MS/MS method for the measurement of insecticide degradates in baby food.
HPLC-MS/MS method for the measurement of insecticide degradates in baby food.
Journal of agricultural and food chemistry (2014-06-10)
Samantha A Radford, Parinya Panuwet, Ronald E Hunter, Dana Boyd Barr, P Barry Ryan
ABSTRACT
A solid phase extraction method was developed to isolate four insecticide degradates from baby food that were measured subsequently using high-performance liquid chromatography-tandem mass spectrometry. The degradates [parent insecticide] measured were malathion dicarboxylic acid [malathion], 3,5,6-trichloro-2-pyridinol [chlorpyrifos, chlorpyrifos methyl] (TCPy), cis/trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropane-1-carboxylic acid [permethrin, cypermethrin, cyfluthrin], and 3-phenoxybenzoic acid [general pyrethroid]. All degradates produced recoveries between 80 and 120% except TCPy in fruit (122% recovery), and all relative standard deviations were <16%. Use of this method demonstrated that insecticide degradates were found in baby foods frequently purchased in the United States, supporting the need for this method. These data will assist in differentiating whether biomarker levels of insecticide metabolites are the result of exposures to the toxic insecticide or its preformed degradate.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%