Merck
  • Capillary electrophoresis-mass spectrometry as a new approach to analyze neonicotinoid insecticides.
Capillary electrophoresis-mass spectrometry as a new approach to analyze neonicotinoid insecticides.
Journal of chromatography. A (2014-08-03)
Laura Sánchez-Hernández, Deamelys Hernández-Domínguez, José Bernal, Christian Neusüß, María T Martín, José L Bernal
ABSTRACT
This paper represents the first report of a capillary electrophoresis (CE) method compatible with mass spectrometry (MS) detection for simultaneously analyzing seven neonicotinoid insecticides (acetamiprid, clothianidin, dinotefuran, imidacloprid, nitenpyram, thiacloprid and thiamethoxam). Different variables affecting CE separation (buffer concentration, pH, applied voltage and injection time) and MS detection (electrospray parameters) were studied. Low limits of detection (LOD) and quantification (LOQ) were achieved for all analytes, ranging from 1.0 to 2.3μg/L, and from 3.5 to 7.2μg/L, respectively. In addition, the proposed method showed itself to be linear in the range from LOQ to 1000μg/L and to be precise, as the relative standard deviations of the migration times were lower than 4% in all cases. Finally, the proposed CE-MS method was applied to assess the efficacy of a beeswax cleaning treatment with oxalic acid to remove residues of three of the most commonly used neonicotinoids (clothianidin, imidacloprid and thiamethoxam), use of which has recently been restricted by the European Union.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Acetic acid, analytical standard
Supelco
Methanol, analytical standard
Supelco
Thiacloprid, PESTANAL®, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%