Merck
Cell dipole behaviour revealed by ECM sub-cellular geometry.
Nature communications (2014-12-11)
Kalpana Mandal, Irène Wang, Elisa Vitiello, Laura Andreina Chacòn Orellana, Martial Balland
ABSTRACT
Cells sense and respond to their mechanical environment by exerting forces on their surroundings. The way forces are modulated by extra-cellular matrix (ECM) properties plays a key role in tissue homoeostasis. Using highly resolved micropatterns that constrain cells into the same square envelope but vary the adhesive geometry, here we investigate how the adhesive micro-environment affects the architecture of actin cytoskeleton and the orientation of traction forces. Our data demonstrate that local adhesive changes can trigger orientational ordering of stress fibres throughout the cell, suggesting that cells are capable of integrating information on ECM geometry at the whole-cell level. Finally, we show that cells tend to generate highly polarized force pattern, that is, unidirectional pinching, in response to adequate adhesive conditions. Hence, the geometry of adhesive environment can induce cellular orientation, a process which may have significant implications for the formation and mechanical properties of tissues.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Fluorescein, European Pharmacopoeia (EP) Reference Standard
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, purum, ≥98.0% (AT)
SAFC
L-Glutamine
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)