Merck
  • Proteomics unravels extracellular vesicles as carriers of classical cytoplasmic proteins in Candida albicans.
Proteomics unravels extracellular vesicles as carriers of classical cytoplasmic proteins in Candida albicans.
Journal of proteome research (2014-11-05)
Ana Gil-Bona, Arancha Llama-Palacios, Claudia Marcela Parra, Fernando Vivanco, César Nombela, Lucía Monteoliva, Concha Gil
ABSTRACT
The commensal fungus Candida albicans secretes a considerable number of proteins and, as in different fungal pathogens, extracellular vesicles (EVs) have also been observed. Our report contains the first proteomic analysis of EVs in C. albicans and a comparative proteomic study of the soluble secreted proteins. With this purpose, cell-free culture supernatants from C. albicans were separated into EVs and EV-free supernatant and analyzed by LC-MS/MS. A total of 96 proteins were identified including 75 and 61 proteins in EVs and EV-free supernatant, respectively. Out of these, 40 proteins were found in secretome by proteomic analysis for the first time. The soluble proteins were enriched in cell wall and secreted pathogenesis related proteins. Interestingly, more than 90% of these EV-free supernatant proteins were classical secretory proteins with predicted N-terminal signal peptide, whereas all the leaderless proteins involved in metabolism, including some moonlighting proteins, or in the exocytosis and endocytosis process were exclusively cargo of the EVs. We propose a model of the different mechanisms used by C. albicans secreted proteins to reach the extracellular medium. Furthermore, we tested the potential of the Bgl2 protein, identified in vesicles and EV-free supernatant, to protect against a systemic candidiasis in a murine model.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology