Merck
  • Up-regulation of glutamine synthesis in microglia activated with endotoxin.
Up-regulation of glutamine synthesis in microglia activated with endotoxin.
Neuroscience letters (2015-02-15)
Kazuyuki Nakajima, Tomoyuki Kanamatsu, Yosuke Takezawa, Shinichi Kohsaka
ABSTRACT
We previously verified that newborn rat brain-derived microglia have the ability to uptake (14)C-glutamate (Glu) through glutamate transporter-1. A given amount of Glu incorporated into microglia was suspected to be metabolized to glutamine (Gln). However, the ability of microglia to do this had not been demonstrated. Thus, in the present study we examined the possibility that primary rat microglia metabolize Glu into Gln. Immunocytochemical and immunoblotting studies indicated that the microglia express glutamine synthetase (GS) protein. As expected from these results, GS activity was actually detected in microglia, although the specific activity was lower than that of astrocytes. Considering this microglial property, it seemed possible that the taken Glu is metabolized to Gln in the cells. To investigate this possibility, we exposed microglia to [(13)C]Glu-containing medium and analyzed the change of Glu to Gln in a nuclear magnetic resonance examination. The results clarified that non-stimulated microglia hardly changed Glu to Gln, but when stimulated with lipopolysaccharide the microglia significantly metabolized [(13)C]Glu to [(13)C]Gln. Microglia were thus, strongly suggested to metabolize Glu to Gln via GS activity when activated in the inflammatory/pathological state of the nervous system.
Product Description
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)