Merck
  • Beneficial effects of soy milk and fiber on high cholesterol diet-induced alteration of gut microbiota and inflammatory gene expression in rats.
Beneficial effects of soy milk and fiber on high cholesterol diet-induced alteration of gut microbiota and inflammatory gene expression in rats.
Food & function (2014-12-06)
Seung-Min Lee, Hye Won Han, Seung Yun Yim
ABSTRACT
We sought to evaluate whether a soy milk and fiber mixture could improve high cholesterol diet-induced changes in gut microbiota and inflammation. Sprague-Dawley rats were administered four different diets: CTRL (AIN76A diet), CHOL (AIN76A with 1% (w/w) cholesterol), SOY (CHOL diet, 20% of which was substituted with freeze-dried soy milk), or S.FIBER (SOY diet with 1.2% (w/w) psyllium, 6.2% (w/w) resistant maltodextrin, and 6.2% (w/w) chicory powder). A lipid profile and gene expression analysis demonstrated that SOY and S.FIBER improved the serum HDL-cholesterol and colonic expression levels of genes in tight junction (ZO-1 and occludin) and inflammation-related (IL-1β, IL-10, and Foxp3) proteins. S.FIBER lowered the serum MCP-1 concentration as well. A gut microbial analysis revealed that CHOL increased the ratio of Firmicutes to Bacteroidetes (F/B ratio). SOY increased the F/B ratio due to an increased proportion of Lactobacillus spp. S.FIBER greatly decreased the F/B ratio. Allobaculum spp. and Parabacteroides spp. exhibited a negative correlation with colonic expression of anti-inflammatory genes such as Foxp3, IL-10, occludin and ZO-1. CHOL increased the relative proportions of Allobaculum spp. and Parabacteroides spp. in the gut, while SOY and S.FIBER decreased these proportions. Diets containing soy milk and fiber mixtures could be beneficial by limiting CHOL-induced colonic inflammation and rescuing CHOL-disturbed gut microbiota.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
SAFC
Sodium deoxycholate