Merck
  • Ataxia telangiectasia mutated inhibits oxidative stress-induced apoptosis by regulating heme oxygenase-1 expression.
Ataxia telangiectasia mutated inhibits oxidative stress-induced apoptosis by regulating heme oxygenase-1 expression.
The international journal of biochemistry & cell biology (2015-01-17)
Ji Hoon Yu, Soon Ok Cho, Joo Weon Lim, Nanhee Kim, Hyeyoung Kim
ABSTRACT
Ataxia telangiectasia (AT) is caused by mutational inactivation of the ataxia telangiectasia mutated (Atm) gene, which is involved in DNA repair. Increased oxidative stress has been shown in human AT cells and neuronal tissues of Atm-deficient mice. Heme oxygenase-1 (HO-1) is an inducible antioxidant enzyme and protects cells against oxidative stress. The purpose of this study is to determine whether ATM induces antioxidant enzyme HO-1 and protects cells from oxidative stress-mediated apoptosis by driving the activation of PKC-δ and NF-κB, by increasing cell viability, and by downregulating DNA fragmentation and apoptotic indicators (apoptosis-inducing factor and cleaved caspase-3). AT fibroblasts stably transfected with human full-length ATM cDNA (YZ5 cells) or the empty vector (MOCK cells) were treated with H2O2 as a source of reactive oxygen species (ROS). As a result, transfection with ATM inhibited ROS-induced cell death and DNA fragmentation in MOCK cells. Transfection with ATM induced expression of HO-1 which was mediated by PKC-δ and NF-κB in H2O2-treated MOCK cells. ZnPP, an HO-1 inhibitor, and transfection with HO-1 siRNA increased ROS levels and apoptosis, whereas hemin, an HO-1 activator, reduced ROS levels and apoptosis in H2O2-treated YZ5 cells. Rottlerin, a PKC-δ inhibitor, inhibited NF-κB activation and HO-1 expression in H2O2-treated YZ5 cells. MOCK cells showed increased cell death, DNA fragmentation, and apoptotic indicators compared to YZ5 cells exposed to H2O2. In addition, transfection with p65 siRNA increased ROS levels and DNA fragmentation, but decreased HO-1 protein levels in H2O2-treated YZ5 cells. In conclusion, ATM induces HO-1 expression via activation of PKC-δ and NF-κB and inhibits oxidative stress-induced apoptosis. A loss of HO-1 induction may explain why AT patients are vulnerable to oxidative stress.
Product Description
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes