Merck
  • Establishment of a mouse model for amiodarone-induced liver injury and analyses of its hepatotoxic mechanism.
Establishment of a mouse model for amiodarone-induced liver injury and analyses of its hepatotoxic mechanism.
Journal of applied toxicology : JAT (2015-04-23)
Shohei Takai, Shingo Oda, Koichi Tsuneyama, Tatsuki Fukami, Miki Nakajima, Tsuyoshi Yokoi
ABSTRACT
Drug-induced liver injury (DILI) is the most frequent cause of post-marketing warnings and withdrawals. Amiodarone (AMD), an antiarrhythmic, presents a risk of liver injury in humans, and its metabolites, formed by cytochrome P450 3A4, are likely more toxic to hepatocytes than AMD is. However, it remains to be clarified whether the metabolic activation of AMD is involved in liver injury in vivo. In this study, to elucidate the underlying mechanisms of AMD-induced liver injury, mice were administered AMD [1000 mg kg(-1), per os (p.o.)] after pretreatment with dexamethasone [DEX, 60 mg kg(-1), intraperitoneal (i.p.)], which induces P450 expression, once daily for 3 days. The plasma alanine aminotransferase (ALT) levels were significantly increased by AMD administration in the DEX-pretreated mice, and the liver concentrations of desethylamiodarone (DEA), a major metabolite of AMD, were correlated with the changes in the plasma ALT levels. Cytochrome c release into the hepatic cytosol and triglyceride levels in the plasma were increased in DEX plus AMD-administered mice. Furthermore, the ratio of reduced glutathione to oxidized glutathione disulfide in the liver significantly decreased in the DEX plus AMD-administered mice. The increase of ALT levels was suppressed by treatment with gadolinium chloride (GdCl3 ), which is an inhibitor of Kupffer cell function. From these results, it is suggested that AMD and/or DEA contribute to the pathogenesis of AMD-induced liver injury by producing mitochondrial and oxidative stress and Kupffer cell activation. This study proposes the mechanisms of AMD-induced liver injury using an in vivo mouse model.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Gadolinium(III) chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) chloride solution, suitable for NMR (reference standard), 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride solution, suitable for NMR (reference standard), 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Sigma-Aldrich
1-Aminobenzotriazole
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration), meets EP testing specifications
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer