Merck
  • Separation and purification of the antioxidant compound hispidin from mushrooms by molecularly imprinted polymer.
Separation and purification of the antioxidant compound hispidin from mushrooms by molecularly imprinted polymer.
Applied microbiology and biotechnology (2015-03-20)
Ning Li, Lijie Zhao, Tzi Bun Ng, Jack Ho Wong, Yan Yan, Zhong Shi, Fang Liu
ABSTRACT
A new molecularly imprinted polymer (MIP), prepared by hispidin as the template molecule, was synthesized and applied as an adsorbent phase for solid phase extraction (SPE) to isolate and enrich hispidin from eight species of mushrooms. The optimization of synthesis and the adsorption behaviors of the MIPs were investigated in detail. In comparison with C18-SPE, MIP-SPE displayed high selectivity and good affinity for hispidin for extract of Phellinus igniarius. The antioxidant activity of the extracts after using the MIPs was evaluated by free radical scavenging activity, and inhibition of erythrocyte hemolysis, and lipid peroxidation. This developed method provided a rapid, selective, and effective approach for separation and enrichment of active compounds from the natural products.
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
Methyl acetoacetate, Arxada quality, ≥99% (GC)
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%