Merck
  • Antioxidant Supplementation Modulates Age-Related Placental Bed Morphology and Reproductive Outcome in Mice.
Antioxidant Supplementation Modulates Age-Related Placental Bed Morphology and Reproductive Outcome in Mice.
Biology of reproduction (2015-07-17)
Elisabete Silva, Ana Isabel Soares, Filipe Costa, José Pedro Castro, Liliana Matos, Henrique Almeida
ABSTRACT
The number of women who delay their first childbirth is increasing. This demographic shift is an important health issue because advanced maternal age is a risk factor for reproductive capacity loss and the occurrence of placental bed disorders that may lead to placenta abruption, preeclampsia, and placenta insufficiency. A redox imbalance status, resulting from the enhanced production of reactive oxygen species or their deficient neutralization, is proposed to occur in this setting. Thus, uterine redox status was evaluated in young (8- to 12-wk-old) and reproductively aged (38- to 42-wk-old) mice. In addition, it was hypothesized that specific dietary antioxidant supplementation would restore the balance and improve the reproductive outcome of aging female mice. To test this hypothesis, two different antioxidants, the nicotinamide adenine dinucleotide phosphate oxidase (NOX) inhibitor apocynin and the superoxide dismutase mimetic 4-hydroxy-2,2,6,6-tetramethylpiperidinyloxy (TEMPOL), were added to the drinking water of female mice prior to and during pregnancy. Compared to younger females, uteri from reproductively aged nonpregnant mice exhibited areas of endometrial cystic dilation, increased level of NOX1 expression, and enhanced protein carbonylation, especially in the apical surface of the luminal epithelium. Both antioxidants decreased protein carbonylation level in the uterus of reproductively aged mice. When reproductively aged females became pregnant, the litter size was smaller and fetuses were heavier. The change was accompanied by a significant decrease in decidua thickness. Provision of apocynin significantly increased litter size and restored decidua thickness. Reproductively aged mice provided with TEMPOL did not evidence such benefits, but whereas apocynin normalized fetal birth weight, TEMPOL further increased it. These findings emphasize that uterine redox balance is important for reproductive success and suggest that age-related redox imbalance might be compensated by specific antioxidant supplementation.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Ponceau S, Dye content 75 %
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Ponceau S solution, BioReagent, suitable (for use in cellulose acetate electrophoresis), 0.1 % (w/v) in 5% acetic acid
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Ponceau S, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)