Merck
  • Opioid receptor delta as a global modulator of skin differentiation and barrier function repair.
Opioid receptor delta as a global modulator of skin differentiation and barrier function repair.
International journal of cosmetic science (2015-02-11)
H Chajra, B Amstutz, K Schweikert, D Auriol, G Redziniak, F Lefèvre
ABSTRACT
The aims of this study were to confirm the properties of selective agonist peptide (Rubixyl) contained in the spinach towards opioid receptor delta. In fact, agonist properties of both spinach peptides (Rubiscolin-5 and Rubixyl) towards opioid receptor delta were demonstrated by Zang et al., but their effects on the other opioid receptors were not studied [1]. We also studied the expression of opioid receptor delta in epidermis under normal and stress condition (inflammatory) and its role in epidermis homeostasis under stress condition in vitro and in vivo. Agonist properties studies were performed using functional agonist cellular model containing human opioid receptors. Opioid receptor delta expression and epidermis homeostasis were studied on human reconstructed epidermis under normal and stress conditions (inflammatory stress) using gene expression (RT-qPCR) and protein expression analysis (immunohistological analysis). Skin repair properties of opioid receptor delta agonist were based on the following parameters TEWL (trans epidermal water loss, hydration and wrinkle depth at periocular and perilabial area) on human volunteers having either intrinsic ageing (more than 40 years old and non-smoker group) and both intrinsic ageing and extrinsic ageing (more than 40 years old and smoker group). We have demonstrated that the Rubixyl peptide is a specific agonist of opioid receptor delta. We have demonstrated that opioid receptor delta expression is modulated under inflammatory condition. The agonist Rubixyl was able to block the depletion of opioid receptor delta seen under inflammatory condition in reconstructed human epidermis. Inflammatory conditions lead to the unbalanced gene and protein expressions of markers involved in epidermis integrity and barrier function properties. The treatment of human reconstructed epidermis with the agonist Rubixyl leads to the normalization of unbalanced gene and protein expressions. In vivo study has confirmed the efficiency of the agonist Rubixyl to repair damaged skin by decreasing TEWL, increasing hydration and decreasing wrinkle depth at the periocular and perilabial area. In this research, we have demonstrated in vitro (on inflamed reconstructed human epidermis, RHE) and in vivo (on human aged volunteers) that activation by natural agonist peptide of opioid receptor delta reduces the skin inflammation thus leading to improvement in epidermis differentiation and skin barrier properties.
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
[D-Pen2,5]-Enkephalin hydrate, ≥95% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
(±)-trans-U-50488 methanesulfonate salt
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%