Merck
  • Improvement of development of equine preantral follicles after 6 days of in vitro culture with ascorbic acid supplementation.
Improvement of development of equine preantral follicles after 6 days of in vitro culture with ascorbic acid supplementation.
Theriogenology (2015-06-16)
R G Gomes, L A Lisboa, C B Silva, M C Max, P C Marino, R L Oliveira, S M González, T R R Barreiros, L S R Marinho, M M Seneda
ABSTRACT
The aim of this study was to evaluate the effects of different concentrations of ascorbic acid (25, 50, and 100 μg/mL) in supplemented minimum essential medium (MEM+) on the development of equine preantral follicles that were cultured in vitro for 2 or 6 days. The contralateral ovaries (n = 5) from five mares in seasonal anestrus were collected from a local abattoir. Nine ovarian tissue fragments of approximately 5 × 5 × 1 mm were obtained from each animal. One fragment was immediately fixed and subjected to histologic analysis (control group; Day 0), and the other eight were placed in PBS supplemented with penicillin (200 IU/mL) and streptomycin (200 mg/mL) at 4 °C for 1 hour (during transport to the laboratory). The fragments were cultured in situ for 2 days (D2) or 6 days (D6) in MEM+ or MEM+ plus ascorbic acid at three different concentrations, establishing the following nine groups: control; MEM+ (D2); MEM+ (D6); MEM+ 25 μg/mL of ascorbic acid (D2); MEM+ 25 μg/mL of ascorbic acid (D6); MEM+ 50 μg/mL of ascorbic acid (D2); MEM+ 50 μg/mL of ascorbic acid (D6); MEM+ 100 μg/mL of ascorbic acid (D2); and MEM+ 100 μg/mL of ascorbic acid (D6). The preantral follicles were classified according to their stage (primordial, primary, secondary, or antral) and their morphology (normal or abnormal). Slides (n = 951) including 4450 histologic sections were evaluated. Follicles were observed in only 4.85% (216 of 4450) of the histologic sections. Of the 407 follicles evaluated, 120 were in the primordial stage and 287 were in different developmental stages; additionally, 43.5% were morphologically normal. After 6 days of culture, the groups cultured with 50 and 100 μg/mL of ascorbic acid differed in terms of follicular development compared with the other groups. On the basis of occurrence of follicular development and the presence of viable follicles, it can be concluded that a positive effect of culture for 6 days in MEM+ supplemented with 50 and 100 μg/mL of ascorbic acid was observed on equine ovarian fragments.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline