Merck
  • Negative and positive temperature dependence of potassium leak in MscS mutants: Implications for understanding thermosensitive channels.
Negative and positive temperature dependence of potassium leak in MscS mutants: Implications for understanding thermosensitive channels.
Biochimica et biophysica acta (2015-05-11)
Piotr Koprowski, Malgorzata A Sliwinska, Andrzej Kubalski
ABSTRACT
Bacterial mechanosensitive channel of small conductance (MscS) is a protein, whose activity is modulated by membrane tension, voltage and cytoplasmic crowding. MscS is a homoheptamer and each monomer consists of three transmembrane helices (TM1-3). Hydrophobic pore of the channel is made of TM3s surrounded by peripheral TM1/2s. MscS gating is a complex process, which involves opening and inactivation in response to the increase of membrane tension. A number of MscS mutants were isolated. Among them mutants affecting gating have been found including gain-of-function (GOF) and loss-of-function (LOF) that open at lower or at higher thresholds, respectively. Previously, using an in vivo screen we isolated multiple MscS mutants that leak potassium and some of them were GOF or LOF. Here we show that for a subset of these mutants K+ leak is negatively (NTD) or positively (PTD) temperature dependent. We show that temperature reliance of these mutants does not depend on how MS gating is affected by a particular mutation. Instead, we argue that NTD or PTD leak is due to the opposite allosteric coupling of the structures that determine the temperature dependence to the channel gate. In PTD mutants an increased hydration of the pore vestibule is directly coupled to the increase in the channel conductance. In NTD mutants, at higher temperatures an increased hydration of peripheral structures leads to complete separation of TM3 and a pore collapse.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)