Merck
  • The bisphosphonate zoledronic acid effectively targets lung cancer cells by inhibition of protein prenylation.
The bisphosphonate zoledronic acid effectively targets lung cancer cells by inhibition of protein prenylation.
Biochemical and biophysical research communications (2015-10-27)
Fan Xie, Pengcheng Li, Jianhua Gong, Jiahong Zhang, Jingping Ma
ABSTRACT
Aberrant activation of oncoproteins such as members of the Ras family is common in human lung cancers. The proper function of Ras largely depends on a post-translational modification termed prenylation. Bisphosphonates have been shown to inhibit prenylation in cancer cells. In this study, we show that zoledronic acid, a third generation bisphosphonate, is effective in targeting lung cancer cells. This is achieved by the induction of apoptosis and inhibition of proliferation, through suppressing the activation of downstream Ras and EGFR signalling by zoledronic acid. The combination of zoledronic acid and paclitaxel or cisplatin (commonly used chemotherapeutic drugs for lung cancer) augmented the activity of either drug alone in in vitro lung cancer cellular system and in vivo lung xenograft mouse model. Importantly, zoledronic acid inhibits protein prenylation as shown by the increased levels of unprenylated Ras and Rap1A. In addition, the effects of zoledronic acid were reversed in the presence of geranylgeraniol and farnesol, further confirming that mechanism of zoledroinc acid's action in lung cancer cells is through prenylation inhibition. Since zoledronic acid is already available for clinic use, these results suggest that it may be an effective addition to the armamentarium of drugs for the treatment of lung cancer.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline