Merck
  • Ascorbic acid provides protection for human chondrocytes against oxidative stress.
Ascorbic acid provides protection for human chondrocytes against oxidative stress.
Molecular medicine reports (2015-08-25)
Zhiqiang Chang, Lifeng Huo, Pengfei Li, Yimin Wu, Pei Zhang
ABSTRACT
Oxidative stress is considered to be an important cause of dysfunction in chondrocytes and articular cartilage degradation, which leads to the pathogenesis of osteoarthritis (OA) and cartilage aging. The present study aimed to assess the effects of the widely applied antioxidant, ascorbic acid (AA), on human chondrocytes against hydrogen peroxide (H2O2) in vitro. Using annexin V‑fluorescein isothiocyanate, 3‑(4,5‑dimethylthiazol‑2‑yl)‑2,5‑diphenyl tetrazolium bromide and senescence‑associated β‑galactosidase assays, the present study identified that AA reduced apoptosis, reduced the loss of viability and markedly decreased H2O2‑mediated senescence in cells treated with H2O2. Furthermore, AA not only stimulated the expression levels of collagens and proteoglycans, but also inhibited the differentiation of chondrocytes under conditions of oxidative stress. In addition, reverse transcription‑quantitative polymerase chain reaction and western blotting demonstrated that AA decreased the activity of nrf2, NF‑κB, AP1 and matrix metalloproteinase‑3, which is stimulated by H2O2. In conclusion, AA efficiently protected human chondrocytes against damage induced by H2O2 by regulating multiple regulatory pathways.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Anti-NRF2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution