Merck
  • Synthesis of cadmium, lead and copper alginate nanobeads as immunosensing probes for the detection of AFP, CEA and PSA.
Synthesis of cadmium, lead and copper alginate nanobeads as immunosensing probes for the detection of AFP, CEA and PSA.
Biosensors & bioelectronics (2015-03-24)
Zifeng Wang, Na Liu, Feng Feng, Zhanfang Ma
ABSTRACT
A double-water-in-oil-emulsion procedure was designed to synthesize cadmium, lead and copper alginate nanobeads less than 200n m diameter under mild conditions. The cadmium, lead and copper alginate nanobeads can be activated to immobilize biomacromolecules and can directly produce distinctive electrochemical signals. Using the novel alginate nanobeads labeled with antibodies as electrochemical probes, a sandwich-type immunosensor was constructed using AFP, CEA and PSA as model analytes. This proposed immunosensor shows wide linear range with detection limits of 0.01, 0.0086 and 0.0075 ng mL(-1) for AFP, CEA and PSA, respectively. Analysis of clinical serum samples using this immunosensor was well consistent with the data determined by the enzyme-linked immunosorbent assay (ELISA). It suggested that the alginate nanobeads electrochemical probes could be generally extended to other multiple analytes detection.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Cadmium chloride
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis