Merck
  • Effects of corn-based reduced-starch diets using alternative carbohydrate sources on performance of lactating Holstein cows.
Effects of corn-based reduced-starch diets using alternative carbohydrate sources on performance of lactating Holstein cows.
Journal of dairy science (2015-04-22)
H M Dann, S M Fredin, K W Cotanch, R J Grant, C Kokko, P Ji, K Fujita
ABSTRACT
Increases in grain prices have led to renewed interest in feeding reduced-starch diets to lactating dairy cows. An experiment was conducted to determine the effects of altering carbohydrate sources and reducing dietary starch on lactational performance, feeding behavior, and ruminal measures of Holstein dairy cows. Fifteen multiparous cows (6 ruminally cannulated) were blocked and assigned to 1 of 5 squares and used in a replicated 3×3 Latin square design with 21-d periods. Cows were fed 1 of 3 experimental diets: a control diet containing 20% brown midrib corn silage, 20% conventional corn silage, and 10% hay crop silage (CON); a reduced-starch high-forage diet containing 53% brown midrib corn silage and 10% hay crop silage (HFOR); and a reduced-starch diet containing the same forages as CON with partial replacement of corn meal by nonforage fiber sources (HNFFS). The CON diet contained (% of dry matter) 26.0% starch and 34.7% neutral detergent fiber (NDF), whereas the HFOR and HNFFS diets contained 21.4 or 21.3% starch and 38.3 or 38.0% NDF, respectively. Dry matter intake tended to be greater for cows fed the CON diet (28.2 kg/d) compared with those fed the HFOR diet (27.2 kg/d). Dry matter intake for cows fed the HNFFS diet was intermediate (27.7 kg/d). Milk yield was greater for cows fed the CON diet (51.6 kg/d) compared with those fed the HFOR diet (48.4 kg/d), but milk fat content tended to increase for cows fed the HFOR diet (3.98%) compared with those fed the CON diet (3.66%). Consequently, fat-corrected and solids-corrected milk yields were unaffected by dietary treatments. Total chewing, eating, and rumination times were similar across all dietary treatments. Rumination time per kilogram of DM was greatest for the HFOR diet, intermediate for the HNFFS diet, and least for the CON diet, whereas rumination time per kilogram of NDF was greatest for the CON diet and least for the HNFFS diet. Mean ruminal pH, NH3-N (mg/dL), and total volatile fatty acid concentrations (mM) were similar across all dietary treatments. Molar proportion of ruminal acetate (mol/100 mol) was increased for cows fed the HFOR diet compared with cows fed the CON diet. Microbial N yield measured by urinary purine derivatives was unaffected by dietary treatment. Reduced-starch diets containing greater amounts of high quality, highly digestible forage or nonforage fiber sources in place of corn meal resulted in similar fat-corrected or solids-corrected milk yield for high-producing dairy cows in the short term.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Molecular Biology, sterile filtered