Merck
  • Application of plastic polymers in remediating wine with elevated alkyl-methoxypyrazine levels.
Application of plastic polymers in remediating wine with elevated alkyl-methoxypyrazine levels.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-04-22)
Andreea Botezatu, Gary J Pickering
ABSTRACT
3-Alkyl-2-methoxypyrazines (MPs) are odour-active compounds that elicit atypical green aromas and flavours in some wines, and are resilient to removal using traditional wine-making approaches. They originate either as contaminants from Coccinellidae beetles inadvertently introduced during wine processing ("ladybug taint") or as grape-derived constituents that are undesirable at elevated levels. In this study we investigated the capacity of a selection of plastic polymers to reduce concentrations of three MPs: isopropyl methoxypyrazine (IPMP), secbutyl methoxypyrazine (SBMP) and isobutyl methoxypyrazine (IBMP). In Trial 1, red wine was spiked with IPMP (20 ng/l), SBMP (20 ng/l) and IBMP (20 ng/l), then separately treated with 13 plastic polymers (surface area 350 cm(2)/l). Three polymers were then identified for further testing based on the results from Trial 1: silicone, ethylene and vinyl acetate (EVA) and a poly-lactic acid-based biodegradable polymer. In Trial 2, the efficacy of these selected polymers to reduce MP levels in red wine was tested as a function of contact time. Solid-phase micro-extraction multi-dimensional GC-MS was used to measure MP levels before and after treatment with the polymers. Results showed significant reductions in all target odorants after 24 h treatment: silicone reduced IPMP and IBMP by 96% and 100%, respectively, while the biodegradable polymer decreased IPMP and IBMP concentrations by 52% and 36%, respectively. EVA was less effective in lowering MP levels (7% IPMP and 23% IBMP after 24 h). Taken overall, the data suggest the potential for the use of poly-lactic acid and silicone in treating wines contaminated by ladybug taint, as well as in reducing high levels of grape-derived MPs.
Product Description
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub