Merck
Flagellin-induced NLRC4 phosphorylation primes the inflammasome for activation by NAIP5.
Proceedings of the National Academy of Sciences of the United States of America (2015-01-22)
Magdalena Matusiak, Nina Van Opdenbosch, Lieselotte Vande Walle, Jean-Claude Sirard, Thirumala-Devi Kanneganti, Mohamed Lamkanfi
ZUSAMMENFASSUNG
The Nlrc4 inflammasome contributes to immunity against intracellular pathogens that express flagellin and type III secretion systems, and activating mutations in NLRC4 cause autoinflammation in patients. Both Naip5 and phosphorylation of Nlrc4 at Ser533 are required for flagellin-induced inflammasome activation, but how these events converge upon inflammasome activation is not known. Here, we showed that Nlrc4 phosphorylation occurs independently of Naip5 detection of flagellin because Naip5 deletion in macrophages abolished caspase-1 activation, interleukin (IL)-1β secretion, and pyroptosis, but not Nlrc4 phosphorylation by cytosolic flagellin of Salmonella Typhimurium and Yersinia enterocolitica. ASC speck formation and caspase-1 expression also were dispensable for Nlrc4 phosphorylation. Interestingly, Helicobacter pylori flagellin triggered robust Nlrc4 phosphorylation, but failed to elicit caspase-1 maturation, IL-1β secretion, and pyroptosis, suggesting that it retained Nlrc4 Ser533 phosphorylating-activity despite escaping Naip5 detection. In agreement, the flagellin D0 domain was required and sufficient for Nlrc4 phosphorylation, whereas deletion of the S. Typhimurium flagellin carboxy-terminus prevented caspase-1 maturation only. Collectively, this work suggests a biphasic activation mechanism for the Nlrc4 inflammasome in which Ser533 phosphorylation prepares Nlrc4 for subsequent activation by the flagellin sensor Naip5.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Roche
cOmplete-ULTRA-Mini-Tabletten, Protease-Inhibitor-Cocktail in EASYpacks , Tablets supplied in foil blister packs.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis