Skip to Content
Merck

Skip To

ATE514316632

AstaTech, Inc.

4-CHLOROISOQUINOLIN-5-OL

Synonym(s):

4-chloroisoquinolin-5-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Ships in 14 days - from Aldrich Partner.
€380.18
250 mg
Ships in 14 days - from Aldrich Partner.
€571.88
1 g
Ships in 14 days - from Aldrich Partner.
€1,362.64

About This Item

Linear Formula:
C9H6O1N1Cl1
CAS Number:
Molecular Weight:
179.61
MDL number:

Vendor SKU: K10066-100MG

€380.18


Ships in 14 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Country of Origin

CN

Inchi Code

1S/C9H6ClNO/c10-7-5-11-4-6-2-1-3-8(12)9(6)7/h1-5,12H

Inchi Key

FIWLPLYJHZKYEO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Room temperature

Physical Form

Yellow to Brown Solid



Still not finding the right product?

Explore all of our products under 4-CHLOROISOQUINOLIN-5-OL


hstatements

Precautionary Statements

Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library