Skip to Content
Merck

14-548-M

MSK1 Protein, active, 10 µg

Active, N-terminal His6-tagged recombinant human MSK1 residues 2-end, for use in Kinase Assays.

Sign In to View Organizational & Contract Pricing

Select a Size

14-548

€601.00

€601.00


Check Cart for Availability

Request a Bulk Order

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 94 kDa

purified by

(Ni2+/NTA-agarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

accession no.

 (FUNCTION: SwissProt: O75582 # Serine/threonine kinase required for the mitogen or stress-induced phosphorylation of the transcription factors CREB (cAMP response element-binding protein) and ATF1 (activating transcription factor-1). Essential role in the control of RELA transcriptional activity in response to TNF. Directly represses transcription via phosphorylation of ′Ser-1′ of histone H2A. Phosphorylates ′Ser-10′ of histone H3 in response to mitogenics, stress stimuli and epidemal growth-factor (EGF), which results in the transcriptional activation of several immediate early genes, including proto-oncogenes c-fos/FOS and c-jun/JUN. May also phosphorylate ′Ser-28′ of histone H3. Mediates the mitogen- and stress-induced phosphorylation of high mobility group protein 14 (HMG-14). )
NM_004755.2

NCBI accession no.

UniProt accession no.

relevant disease(s)

metabolic disorders; cardiovascular diseases

Gene Information

human ... RPS6KA5(9252)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
14-616-M14-624-M14-452-M
biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

technique(s)

activity assay: suitable (kinase)

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 94 kDa

mol wt

Mw 89.9 kDa

mol wt

Mw 57 kDa

mol wt

Mw 59 kDa

Gene Information

human ... RPS6KA5(9252)

Gene Information

human ... RPS6KA4(8986)

Gene Information

human ... STK4(6789)

Gene Information

human ... PDPK1(5170)

UniProt accession no.

O75582

UniProt accession no.

O75676

UniProt accession no.

Q13043

UniProt accession no.

O15530

General description

MSK1
N-terminal His6-tagged recombinant human MSK1 residues 2-end

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: MSK1
Target Sub-Family: AGC

Packaging

Also available in 10 μg size and in bulk
Also available in 250 µg size --call for pricing and availability and reference catalog number 14-548M when ordering the 250 µg size.

Physical form

Ni2+/NTA-agarose

Preparation Note

6 months at -20°C

Analysis Note

routinely evaluated by phosphorylation of Crosstide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judit M Pérez Ortiz et al.
Neurobiology of disease, 116, 93-105 (2018-05-15)
Spinocerebellar ataxia type 1 (SCA1) is a polyglutamine (polyQ) repeat neurodegenerative disease in which a primary site of pathogenesis are cerebellar Purkinje cells. In addition to polyQ expansion of ataxin-1 protein (ATXN1), phosphorylation of ATXN1 at the serine 776 residue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service