Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
UNSPSC Code:
41116107
Skip To
grade
certified reference material
Agency
ERM®
manufacturer/tradename
JRC
application(s)
food and beverages
format
matrix material
storage temp.
2-8°C
General description
For more information please see: EURM022
Legal Information
ERM is a registered trademark of European Commission
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service