Skip to Content
Merck

GF75251909

Palladium

microfoil, disks, 10mm, thinness 0.05μm, specific density 63.6μg/cm2, permanent mylar 3.5μm support, 99.99%

Synonym(s):

Palladium, PD004300

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141733
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Palladium, microfoil, disks, 10mm, thinness 0.05μm, specific density 63.6μg/cm2, permanent mylar 3.5μm support, 99.99%

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 752-519-09

resistivity

9.96 μΩ-cm, 20°C

diam. × thickness

10 mm × 0.05 μm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF26034230GF52930762GF47424528
assay

99.99%

assay

99.99%

assay

99.99%

assay

99.99%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 752-519-09

manufacturer/tradename

Goodfellow 260-342-30

manufacturer/tradename

Goodfellow 529-307-62

manufacturer/tradename

Goodfellow 474-245-28

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of heterocycles via palladium-catalyzed carbonylations.
Xiao-Feng Wu et al.
Chemical reviews, 113(1), 1-35 (2012-10-09)
Amanda J Hickman et al.
Nature, 484(7393), 177-185 (2012-04-14)
Copper and palladium catalysts are critically important in numerous commercial chemical processes. Improvements in the activity, selectivity and scope of these catalysts could drastically reduce the environmental impact, and increase the sustainability, of chemical reactions. One rapidly developing strategy for
Update 1 of: Synthesis and functionalization of indoles through palladium-catalyzed reactions.
Sandro Cacchi et al.
Chemical reviews, 111(5), PR215-PR283 (2011-05-12)
Masahiro Toyota
Natural product communications, 8(7), 999-1004 (2013-08-29)
A novel palladium-catalyzed intramolecular oxidative alkylation of unactivated olefins is described. This protocol was devised to solve one of the drawbacks of the original palladium-catalyzed cycloalkenylation that we developed. We call this new procedure the 'second generation palladium-catalyzed cycloalkenylation'. This
Jana Doháňošová et al.
Molecules (Basel, Switzerland), 18(6), 6173-6192 (2013-05-28)
The palladium (II)-catalysed reactions of alkenols and aminoalkenols such as oxycarbonylations or bicyclisations are powerful methods for the construction of oxygen and nitrogen-containing heterocyclic compounds. This review highlights recent progress in the development of the asymmetric palladium(II)-catalysed Wacker-type cyclisations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service