Skip to Content

BDPH9BCE02CA

Bide Pharmatech

(6-Bromo-2-fluoropyridin-3-yl)methanol

Synonym(s):

(6-bromo-2-fluoro-3-pyridinyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O1N1Br1F1
CAS Number:
Molecular Weight:
206.01
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H5BrFNO/c7-5-2-1-4(3-10)6(8)9-5/h1-2,10H,3H2

Inchi Key

JQIREWOWBXROGC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library