Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Ships in 14 days - from Aldrich Partner.
| 1.741,50 kr. |
250 mg | Ships in 14 days - from Aldrich Partner.
| 2.346,00 kr. |
500 mg | Ships in 14 days - from Aldrich Partner.
| 4.110,75 kr. |
1 g | Ships in 14 days - from Aldrich Partner.
| 5.241,75 kr. |
2.5 g | Ships in 14 days - from Aldrich Partner.
| 9.931,50 kr. |
5 g | Ships in 14 days - from Aldrich Partner.
| 14.543,25 kr. |
10 g | Ships in 14 days - from Aldrich Partner.
| 21.407,25 kr. |
About This Item
Vendor SKU: EN300-141355-100MG
1.741,50 kr.
Ships in 14 days - from Aldrich Partner.
Inchi Code
1S/C6H16N2O/c1-9-6-5-8-4-2-3-7/h8H,2-7H2,1H3
Inchi Key
LCVKUHWARTYVOZ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95
Storage temp.
RT
Country of Origin
UA
Physical Form
liquid
hstatements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

