07-463-S
Anti-trimethyl-Histone H4 (Lys20) Antibody, Trial Size
from rabbit, purified by affinity chromatography
Synonym(s):
H4K20me3, Histone H4 (trimethyl K20), H4 histone family, member A, Histone 1, H4a, histone cluster 1, H4a
Select a Size
About This Item
Skip To
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
rat, human, mouse
technique(s)
ChIP: suitable
dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
trimethylation (Lys20)
Gene Information
human ... H4C1(8359)
mouse ... H4C1(326619)
1 of 4
This Item | 04-079-S | 07-463 | 04-079 |
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| antibody form affinity isolated antibody | antibody form culture supernatant | antibody form affinity isolated antibody | antibody form culture supernatant |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| clone polyclonal | clone monoclonal | clone polyclonal | clone monoclonal |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
General description
Immunogen
Application
Epigenetics & Nuclear Function
Histones
Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides (see tables) were probed with Anti-trimethyl-Histone H4 (Lys20) (4 µg/mL).
Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated Histone H4 using chromatin from HeLa cells.
Immunoprecipitation Analysis: A representative lot immunoprecipitated trimethyl-Histone H4 (Lys20) from HeLa cell lysate.
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
HeLa acid extract and various recombinant Histone proteins.
Western Blot Analysis: 2 µg/mL of this antibody detected trimethyl-Histone H4 (Lys20) in 10 µg of HeLa acid extract.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service

