Select a Size
3.630,00 kr.
3.630,00 kr.
About This Item
Skip To
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
2H5, monoclonal
form
buffered aqueous solution
mol wt
antigen ~42.5 kDa
species reactivity
human
technique(s)
indirect ELISA: suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... SULF2(55959)
1 of 4
This Item | SAB2501213 | MABC586 | SAB1407186 |
|---|---|---|---|
| antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| Gene Information human ... SULF2(55959) | Gene Information human ... SULF2(55959) | Gene Information human ... SULF2(55959) | Gene Information human ... SULF1(23213) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| clone 2H5, monoclonal | clone polyclonal | clone 8G1, monoclonal | clone polyclonal |
| biological source mouse | biological source goat | biological source mouse | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate - | conjugate unconjugated |
General description
Immunogen
Sequence
MPGLTCFTHDNQHWQTAPFWTLGPFCACTSANNNTYWCMRTINETHNFLFCEFATGFLEYFDLNTDPYQLMNAVNTLDRDVLNQLHVQLMELRSCKGYKQCNPRTRNMDLGLKDGGSYEQYRQFQRRKWPEMKRPSSKSLGQLWEGWEG
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


