Merck
  • Metformin promotes irisin release from murine skeletal muscle independently of AMP-activated protein kinase activation.
Metformin promotes irisin release from murine skeletal muscle independently of AMP-activated protein kinase activation.
Acta physiologica (Oxford, England) (2014-11-11)
D-J Li, F Huang, W-J Lu, G-J Jiang, Y-P Deng, F-M Shen
ABSTRACT
Irisin, a novel myocyte-secreted hormone mediating beneficial effects of exercise on metabolism, is supposed to be an ideal therapeutic target for metabolic disorders such as obesity and diabetes. Here, we investigated the potential effects of metformin and glibenclamide, two antidiabetic medicines, on irisin release in mouse. Wild-type and diabetic obese db/db mice were administrated with metformin and glibenclamide for 2 weeks, and cultured C2C12 myotubes were treated by metformin. Expression of irisin precursor FNDC5 was measured and blood irisin concentration was detected. AMP-activated protein kinase (AMPK) was blocked by chemical inhibitor compound C or knocking down with specific siRNA. The mRNA and protein expression of FNDC5 in skeletal muscle and blood irisin concentrations were lower in diabetic db/db mice than those in wild-type mice. Metformin and glibenclamide decreased blood glucose in db/db mice. Metformin, but not glibenclamide, increased intramuscular FNDC5 mRNA/protein expression and blood irisin levels. Additionally, the reductions of blood glucose and body weight in metformin-treated db/db mice were positively associated with blood irisin concentrations. In C2C12 myotubes, metformin upregulated intracellular FDNC5 mRNA/protein expression and promoted irisin release. Although metformin activated AMPK signalling in skeletal muscle cells, disrupting of AMPK signalling by chemical inhibitor or siRNA-mediated knockdown did not abolish the promoting effect of metformin on irisin release. Metformin promotes irisin release from murine skeletal muscle into blood, independently of AMPK pathway activation. Our results suggest that stimulation of irisin may be a novel molecular mechanism of metformin which is widely used for treatment of metabolic disorders.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%