Merck
  • Myostatin is localized in extravillous trophoblast and up-regulates migration.
Myostatin is localized in extravillous trophoblast and up-regulates migration.
The Journal of clinical endocrinology and metabolism (2014-08-06)
Hassendrini N Peiris, Carlos Salomon, Diane Payton, Keith Ashman, Kanchan Vaswani, Anthony Chan, Gregory E Rice, Murray D Mitchell
ABSTRACT
Myostatin is a highly conserved secretory protein that negatively regulates muscle development by affecting both proliferation and differentiation of muscle cells. In human placentae the expression of myostatin is negatively correlated with gestational age, and in placental explants, myostatin acts to facilitate glucose uptake. Myostatin expression is known to be higher in the placentae of pregnancies complicated by preeclampsia. Proper placental development is crucial for a healthy and successful pregnancy. Alterations to the function of the placental cells after treatment with myostatin have not previously been published. This study investigated the localization of myostatin in extravillous trophoblast (EVT) of human placentae. Furthermore, the effect of myostatin treatment on the proliferative and migrative capabilities of these placental cells was investigated. Myostatin is localized in EVT, as identified by the immunohistochemistry of third-trimester placentae and immunocytochemistry of first-trimester EVT isolations positively staining for myostatin and human leukocyte antigen-G. Treatment of an EVT cell line (HTR-8/SVneo) and primary isolated EVT with varied concentrations of myostatin resulted in a significant increase in the proliferation (HTR-8/SVneo; P < .0001) and migration (HTR-8/SVneo and primary isolated EVT; P < .05), with proliferation being dose dependent and migration being dose independent. Myostatin localization was positively identified in EVT. Myostatin positively affected proliferation (HTR-8/SVneo) and migration of EVT (HTR-8/SVneo and primary isolated EVT). For the first time, the effect of myostatin treatment on placental cells is described. The results provide a base from which further in vitro investigations on myostatin's ability to modulate placental cell function can be made.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Anti-VIM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KRT7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, Molecular Biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard