Merck
  • Expression of microRNA-155 correlates positively with the expression of Toll-like receptor 7 and modulates hepatitis B virus via C/EBP-β in hepatocytes.
Expression of microRNA-155 correlates positively with the expression of Toll-like receptor 7 and modulates hepatitis B virus via C/EBP-β in hepatocytes.
Journal of viral hepatitis (2015-02-28)
N Sarkar, R Panigrahi, A Pal, A Biswas, S P Singh, S K Kar, M Bandopadhyay, D Das, D Saha, T Kanda, M Sugiyama, S Chakrabarti, A Banerjee, R Chakravarty
ABSTRACT
Effective recognition of viral infection and successive activation of antiviral innate immune responses are vital for host antiviral defence, which largely depends on multiple regulators, including Toll-like receptors (TLRs) and microRNAs. Several early reports suggest that specific TLR-mediated immune responses can control hepatitis B virus (HBV) replication and express differentially with disease outcome. Considering the versatile function of miR-155 in the TLR-mediated innate immune response, we aimed to study the association between miR-155 and TLRs and their subsequent impact on HBV replication using both a HBV-replicating stable cell line (HepG2.2.15) and HBV-infected liver biopsy and serum samples. Our results showed that miR-155 was suppressed during HBV infection and a subsequent positive correlation of miR-155 with TLR7 activation was noted. Further, ectopic expression of miR-155 in vitro reduced HBV load as evidenced from reduced viral DNA, mRNA and subsequently reduced level of secreted viral antigens (HBsAg and HBeAg). Our results further suggested that CCAAT/enhancer-binding protein-β (C/EBP-β), a positive regulator of HBV transcription, was inhibited by miR-155. Taken together, our study established a correlation between miR-155 and TLR7 during HBV infection and also demonstrated in vitro that increased miR-155 level could help to reduce HBV viral load by targeting C/EBP-β.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O