Merck
  • Wetting effects versus ion pairs diffusivity: interactions of anionic surfactants with highly soluble cationic drugs and its impact on tablet dissolution.
Wetting effects versus ion pairs diffusivity: interactions of anionic surfactants with highly soluble cationic drugs and its impact on tablet dissolution.
Journal of pharmaceutical sciences (2015-05-29)
Divyakant Desai, Benjamin Wong, Yande Huang, Qingmei Ye, Hang Guo, Ming Huang, Peter Timmins
ABSTRACT
A study was conducted to develop a mechanistic understanding of dissolution of a highly soluble cationic drug, metformin hydrochloride, under the influence of anionic surfactants, sodium alkyl sulfates. The surfactants did not influence the saturated solubility of the drug, but reduced the surface tension of the dissolution media as the alkyl chain length increased. Their influence on tablet wetting based on the contact angles did not show any trend. The dissolution of 850 mg metformin hydrochloride tablets in 0.1 N HCl and pH 4.5 acetate buffer with 0.01% (w/v) sodium n-octyl sulfate (C8), sodium n-decyl sulfate (C10), or sodium n-tetradecyl sulfate (C14) was similar to the control, but was enhanced by sodium lauryl sulfate (C12). At 0.1% (w/v) concentration, the dissolution was not enhanced by C12 because the reduction in surface tension was counterbalanced by an increase in hydrophobic ion pairs that showed slower diffusivity by nuclear magnetic resonance. At 0.1% (w/v), metformin also formed an insoluble salt (1:2 molar ratios) with C10 (pH 1.2), C12, and C14 (pH 1.2 and 4.5) but not with C8. Three competing factors influenced the drug dissolution by surfactants: reduction in surface tension of the dissolution media, ion pairs with low diffusivity, and formation of an insoluble salt.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Sodium octyl sulfate, ≥95%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Tetradecyl sulfate sodium salt, 95%
Sigma-Aldrich
Magnesium stearate, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
Magnesium stearate, technical grade
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis