Merck
  • The Stage- and Cell Type-Specific Localization of Fragile X Mental Retardation Protein in Rat Ovaries.
The Stage- and Cell Type-Specific Localization of Fragile X Mental Retardation Protein in Rat Ovaries.
Reproductive sciences (Thousand Oaks, Calif.) (2015-06-04)
Noriyuki Takahashi, Wataru Tarumi, Masanori T Itoh, Bunpei Ishizuka
ABSTRACT
Premutations of the fragile X mental retardation 1 (FMR1) gene are associated with increased risk of primary ovarian insufficiency. Here we examined the localization of the Fmr1 gene protein product, fragile X mental retardation protein (FMRP), in rat ovaries at different stages, including fetus, neonate, and old age. In ovaries dissected from 19 days postcoitum embryos, the germ cells were divided into 2 types: one with decondensed chromatin in the nucleus was FMRP positive in the cytoplasm, but the other with strongly condensed chromatin in the nucleus was FMRP negative in the cytoplasm. The FMRP was predominantly localized to the cytoplasm of oocytes in growing ovarian follicles. Levels of FMRP in oocytes from elderly (9 or 14 months of age) ovaries were lower than in those from younger ovaries. These results suggest that FMRP is associated with the activation of oogenesis and oocyte function. Especially, FMRP is likely to be implicated in germline development during oogenesis.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)