Merck
  • Myosin heavy chain 10 (MYH10) is required for centriole migration during the biogenesis of primary cilia.
Myosin heavy chain 10 (MYH10) is required for centriole migration during the biogenesis of primary cilia.
Biochemical and biophysical research communications (2015-04-18)
Hyowon Hong, Jongshin Kim, Joon Kim
ABSTRACT
The actin cytoskeleton has been implicated in the assembly of cilia, but roles of actin-dependent motor proteins in ciliogenesis remain unclear. Myosin heavy chain 10 (MYH10), one of the isoforms of non-muscle myosin II, is known to mediate centrosome reorientation during cell migration. Here we show that MYH10 is required for centriole migration to the apical plasma membrane, which occurs at the onset of ciliogenesis. Knockdown of MYH10 in RPE1 cells caused a reduction in the levels of cortical filamentous actin (F-actin) and its binding protein EZRIN. Moreover, both centriole migration and subsequent cilium assembly were defective in MYH10 depleted cells. We further found that MYH10 influences centrosomal recruitment of IFT88, which is required for the transport of building blocks to the ciliary tip. The role of MYH10 in IFT88 recruitment appears to be indirect in that there is a correlation between centriolar IFT88 levels and centriolar positions along the apical-basal axis during ciliogenesis. Our results indicate that MYH10 contributes to ciliogenesis in RPE1 cells by promoting cortical actin-dependent centriole migration.
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)