Saltar al contenido
Merck

939218

Protein delivery system

Human SHH, PODS®

Sinónimos:

PODS®

Iniciar sesión para ver los precios por organización y contrato.

Seleccione un Tamaño


Acerca de este artículo

NACRES:
NA.21
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarle

Nombre del producto

Protein delivery system, Human SHH, PODS®

biological source

(Spodoptera frugiperda (Sf9) cell culture)

form

(monodisperse crystals)

packaging

1 ea of 25 μg

technique(s)

: suitable using 3D bioprinting | cell culture

impurities

<0.06 EU/mL Endotoxin

color

colorless

storage temp.

2-8°C

Quality Level

¿Está buscando productos similares? Visita Guía de comparación de productos

Application

PODS are composed of a bioactive cargo protein within a polyhedrin crystal lattice, with polyhedrin as the main protein. The 25 µg SKU size indicates the amount of bioactive cargo, typically 0.5% to 5% of the total protein. For instance, a 25 µg SKU at a 2% cargo loading contains 1.25 mg of total protein. PODS enable sustained release of cargo proteins when proteases degrade the polyhedrin scaffold. Growth factors are not bioavailable until released, and their concentration in media depends on the amount added and the release rate. In 10% serum, peak levels are usually reached within 24-48 hours, with about 20% of the cargo becoming soluble.

PODS often require lower concentrations than conventional growth factors, as they maintain effective levels. Pre-incubating with serum or proteases enhances availability, while conventional factors can be added for immediate effects. Optimal amounts should be determined empirically.

Features and Benefits

  • Protein delivery system, PODS human SHH comes with molecular weight of 25 kDa, length 220 aa and endotoxin level <0.06 EU/ml. PODS co-crystals may be reconstituted at 200 million co-crystals/ml in sterile PBS. 20% glucose has a buoyant density closer to PODS co-crystals and can be useful for aliquoting.
  • This product is produced with no animal derived raw products. All processing and handling employs animal free equipment and animal free protocols. PODS co-crystals are highly stable when stored in aqueous solution (pH range 6 - 8).
  • PODS provides durability, stability and sustained release, addressing the limitations of conventional and enabling researchers to effortlessly control and manipulate cells in 2D and 3D culture as well as in-vivo.

General description

Protein delivery system, SHH, PODS contains the polyhedrin protein co-crystalized with Human SHH. Sonic hedgehog (SHH) is a member of a small group of hedgehog secreted proteins that are essential for development in both vertebrates and invertebrates. There are three mammalian hedgehog homologues, sonic, desert, and indian, that signal via the Patched-1 and Patched-2 receptors. SHH is a morphogen that is essential during vertebrate organogenesis and adult stem cell division.

Legal Information

PODS is a registered trademark of Cell Guidance Systems Ltd

Clase de almacenamiento

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

¿No ve la versión correcta?

Si necesita una versión concreta, puede buscar un certificado específico por el número de lote.

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Gabriela Basile Carballo et al.
Cell communication and signaling : CCS, 16(1), 11-11 (2018-03-22)
Hedgehog (Hh) signaling pathway plays an essential role during vertebrate embryonic development and tumorigenesis. It is already known that Sonic hedgehog (Shh) pathway is important for the evolution of radio and chemo-resistance of several types of tumors. Most of the
Eva Tiecke et al.
Methods in molecular biology (Clifton, N.J.), 397, 23-33 (2007-11-21)
Here, we describe methods for applying Sonic hedgehog (Shh) to developing chick limbs. The Sonic hedgehog gene is expressed in the polarizing region, a signaling region at the posterior margin of the limb bud and application of Shh-expressing cells or
Erwan Bezard et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 17(15), 2337-2338 (2003-10-04)
It is well established that members of the hedgehog family are involved in tissue patterning during development. We herein show that sonic hedgehog signaling molecules are differentially regulated by dopamine depletion in the basal ganglia of adult animals and specifically

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico