Merck
Cdc14 and PP2A Phosphatases Cooperate to Shape Phosphoproteome Dynamics during Mitotic Exit.
Cell reports (2019-11-14)
Sandra A Touati, Lorena Hofbauer, Andrew W Jones, Ambrosius P Snijders, Gavin Kelly, Frank Uhlmann
RESUMEN
Temporal control over protein phosphorylation and dephosphorylation is crucial for accurate chromosome segregation and for completion of the cell division cycle during exit from mitosis. In budding yeast, the Cdc14 phosphatase is thought to be a major regulator at this time, while in higher eukaryotes PP2A phosphatases take a dominant role. Here, we use time-resolved phosphoproteome analysis in budding yeast to evaluate the respective contributions of Cdc14, PP2ACdc55, and PP2ARts1. This reveals an overlapping requirement for all three phosphatases during mitotic progression. Our time-resolved phosphoproteome resource reveals how Cdc14 instructs the sequential pattern of phosphorylation changes, in part through preferential recognition of serine-based cyclin-dependent kinase (Cdk) substrates. PP2ACdc55 and PP2ARts1 in turn exhibit a broad substrate spectrum with some selectivity for phosphothreonines and a role for PP2ARts1 in sustaining Aurora kinase activity. These results illustrate synergy and coordination between phosphatases as they orchestrate phosphoproteome dynamics during mitotic progression.
MATERIALES
Número de producto
Marca
Descripción del producto
Millipore
Nucleasas Benzonase®, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Roche
cOmplete, combinación de inhibidores de proteasas sin EDTA, Tablets provided in EASYpacks
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Indoxyl β-D-glucoside, ≥97%
Sigma-Aldrich
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution