Merck
The effect of epitope density and antibody affinity on the ELISA as analysed by monoclonal antibodies.
Journal of immunological methods (1984-08-03)
A M Lew
RESUMEN
The antibody titre and the optical density values in an ELISA are influenced by the epitope density of the antigen and the affinity of the antibody tested. This has major implications in the interpretation of ELISA results.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
IgG1, Lambda from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)