Merck
Live slow-frozen human tumor tissues viable for 2D, 3D, ex vivo cultures and single-cell RNAseq.
Communications biology (2022-10-29)
Gaetana Restivo, Aizhan Tastanova, Zsolt Balázs, Federica Panebianco, Maren Diepenbruck, Caner Ercan, Bodgan-T Preca, Jürg Hafner, Walter P Weber, Christian Kurzeder, Marcus Vetter, Simone Münst Soysal, Christian Beisel, Mohamed Bentires-Alj, Salvatore Piscuoglio, Michael Krauthammer, Mitchell P Levesque
RESUMEN
Biobanking of surplus human healthy and disease-derived tissues is essential for diagnostics and translational research. An enormous amount of formalin-fixed and paraffin-embedded (FFPE), Tissue-Tek OCT embedded or snap-frozen tissues are preserved in many biobanks worldwide and have been the basis of translational studies. However, their usage is limited to assays that do not require viable cells. The access to intact and viable human material is a prerequisite for translational validation of basic research, for novel therapeutic target discovery, and functional testing. Here we show that surplus tissues from multiple solid human cancers directly slow-frozen after resection can subsequently be used for different types of methods including the establishment of 2D, 3D, and ex vivo cultures as well as single-cell RNA sequencing with similar results when compared to freshly analyzed material.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Seroalbúmina bovina, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Toxina del cólera from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Desoxirribonucleasa I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Seroalbúmina bovina, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Roche
Dispase® II (neutral protease, grade II), lyophilized, from bacterial, Roche, pkg of 5 × 1 g
Sigma-Aldrich
Apo-transferrina human, powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
SB 202190 monohydrochloride hydrate, ≥98% (HPLC)