Merck
  • Comparative effectiveness of pharmacologic interventions for knee osteoarthritis: a systematic review and network meta-analysis.
Comparative effectiveness of pharmacologic interventions for knee osteoarthritis: a systematic review and network meta-analysis.
Annals of internal medicine (2015-01-07)
Raveendhara R Bannuru, Christopher H Schmid, David M Kent, Elizaveta E Vaysbrot, John B Wong, Timothy E McAlindon
RESUMEN
The relative efficacy of available treatments of knee osteoarthritis (OA) must be determined for rational treatment algorithms to be formulated. To examine the efficacy of treatments of primary knee OA using a network meta-analysis design, which estimates relative effects of all treatments against each other. MEDLINE, EMBASE, Web of Science, Google Scholar, Cochrane Central Register of Controlled Trials from inception through 15 August 2014, and unpublished data. Randomized trials of adults with knee OA comparing 2 or more of the following: acetaminophen, diclofenac, ibuprofen, naproxen, celecoxib, intra-articular (IA) corticosteroids, IA hyaluronic acid, oral placebo, and IA placebo. Two reviewers independently abstracted study data and assessed study quality. Standardized mean differences were calculated for pain, function, and stiffness at 3-month follow-up. Network meta-analysis was performed using a Bayesian random-effects model; 137 studies comprising 33,243 participants were identified. For pain, all interventions significantly outperformed oral placebo, with effect sizes from 0.63 (95% credible interval [CrI], 0.39 to 0.88) for the most efficacious treatment (hyaluronic acid) to 0.18 (CrI, 0.04 to 0.33) for the least efficacious treatment (acetaminophen). For function, all interventions except IA corticosteroids were significantly superior to oral placebo. For stiffness, most of the treatments did not significantly differ from one another. Lack of long-term data, inadequate reporting of safety data, possible publication bias, and few head-to-head comparisons. This method allowed comparison of common treatments of knee OA according to their relative efficacy. Intra-articular treatments were superior to nonsteroidal anti-inflammatory drugs, possibly because of the integrated IA placebo effect. Small but robust differences were observed between active treatments. All treatments except acetaminophen showed clinically significant improvement from baseline pain. This information, along with the safety profiles and relative costs of included treatments, will be helpful for individualized patient care decisions. Agency for Healthcare Research and Quality.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
USP
Diclofenac sodium, United States Pharmacopeia (USP) Reference Standard
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Supelco
Ibuprofen
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Diclofenac sodium salt, analytical standard
Sigma-Aldrich
Naproxen, meets USP testing specifications