Merck
In vivo biodistribution and clearance of peptide amphiphile micelles.
Nanomedicine : nanotechnology, biology, and medicine (2014-09-10)
Eun Ji Chung, Laurie B Mlinar, Matthew J Sugimoto, Kathryn Nord, Brian B Roman, Matthew Tirrell
RESUMEN
Peptide amphiphiles (PAs) are promising biomaterials for medical applications. To translate the use of PAs successfully from laboratories to clinics, in vivo studies regarding the safety of these nanomaterials are required. To examine the toxicity and clearance of PA biomaterials, we intravenously administered cy7-labeled, spherical PA micelles, control micelles without a peptide sequence, or PBS in a murine model and investigated biocompatibility, biodistribution, and clearance. Both peptide and non-peptide labeled micelles were approximately 8 nm in diameter, but of opposite surface charge. Neither micelle type caused aggregation or hemolysis of red blood cells. All micelles primarily accumulated in the bladder and were present in urine samples confirming elimination through renal clearance. Ex vivo imaging showed that micelles were also found in the liver suggesting some involvement of the reticuloendothelial system. However, no evidence of toxicity was found within the liver, spleen, kidney, bladder, intestines, lung, and heart. Safety studies related to peptide amphiphile biomaterials are discussed in this paper, demonstrating that organotoxicity is unlikely with these materials, however, RES activation in the liver may be of consideration in further studies and needed for potential applications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido trifluoroacético, ReagentPlus®, 99%
Sigma-Aldrich
Ácido trifluoroacético, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Acetonitrilo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sacarosa, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrilo, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrilo, anhydrous, 99.8%
Sigma-Aldrich
Ácido trifluoroacético, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sacarosa, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Acetonitrilo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido trifluoroacético, ≥99%, for protein sequencing
Supelco
Sacarosa, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sacarosa, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
Sacarosa, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sacarosa, ACS reagent
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Acetonitrilo, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrilo, electronic grade, 99.999% trace metals basis
Supelco
Ácido trifluoroacético, analytical standard
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Acetonitrilo, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitrilo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sacarosa, ≥99% (GC), Grade I, suitable for plant cell culture
Supelco
Acetonitrilo, analytical standard