Merck
  • Sensitive impedimetric biosensor based on duplex-like DNA scaffolds and ordered mesoporous carbon nitride for silver(I) ion detection.
Sensitive impedimetric biosensor based on duplex-like DNA scaffolds and ordered mesoporous carbon nitride for silver(I) ion detection.
The Analyst (2014-10-28)
Yaoyu Zhou, Lin Tang, Xia Xie, Guangming Zeng, Jiajia Wang, Yaocheng Deng, Guide Yang, Chen Zhang, Yi Zhang, Jun Chen
RESUMEN
This study demonstrates a new, unlabeled immobilized DNA-based biosensor with ordered mesoporous carbon nitride material (MCN) for the detection of Ag(+) by electrochemical impedance spectroscopy (EIS) with [Fe(CN)6](4-/3-) as the redox couple. The unlabeled immobilized DNA initially formed the hairpin-like structure through hybridization with the probe, and then changed to duplex-like structure upon interaction with Ag(+) in solution to form a C-Ag(+)-C complex at electrode surface. As a result, the interfacial charge-transfer resistance of the electrode towards the [Fe(CN)6](4-/3-) redox couple was changed. Thus, a declined charge transfer resistance (Rct) was obtained, corresponding to Ag(+) concentration. MCN provide an excellent platform for DNA immobilization and faster electron transfer. Impedance data were analyzed with the help of Randles equivalent circuit. The lower detection limit of the biosensor for Ag(+) is 5 × 10(-11) M with good specificity. All results showed that this novel approach provides a reliable method for Ag(+) detection with sensitivity and specificity, potentially useful for practical applications. Moreover, other DNA detection methods for more heavy metals may be obtained from this idea and applied in the environmental field.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Base Trizma®, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Base Trizma®, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Supelco
Óxido de etileno solution, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
Tris(hidroximetil)-aminometano, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Base Trizma®, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Óxido de etileno solution, 2.5-3.3 M in THF
Sigma-Aldrich
Óxido de (±)-propileno, ReagentPlus®, ≥99%
Sigma-Aldrich
Óxido de (±)-propileno, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Base Trizma®, ≥99.0% (T)
Sigma-Aldrich
Base Trizma®, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Base Trizma®, ≥99.9% (titration), crystalline
Sigma-Aldrich
Base Trizma®, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Base Trizma®, 99.7% (T), puriss. p.a.
Supelco
Óxido de (±)-propileno, analytical standard
Supelco
Base Trizma®, reference material for titrimetry, certified by BAM, >99.5%
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Trometamol, European Pharmacopoeia (EP) Reference Standard
SAFC
Tromethamine