Merck
MILI, a PIWI family protein, inhibits melanoma cell migration through methylation of LINE1.
Biochemical and biophysical research communications (2015-01-15)
Xiuxing Wang, Chen Jiang, Bingyuan Fu, Ruilou Zhu, Fan Diao, Na Xu, Zhong Chen, Weiwei Tao, Chao-Jun Li
RESUMEN
MILI, a member of the PIWI/AGO gene family, has been well documented to maintain genome integrity by transposon silencing in animal germ cells. It has been reported to be selectively expressed in precancerous stem cells (pCSCs), tumor cell lines and various malignancies. However, the underlying mechanism remains largely unclear. Here, we found that MILI is expressed in the melanoma cell line B16 but not in the highly metastatic mouse melanoma model B16BL6. Interestingly, the knockdown of MILI in B16 could activate MAGEA expression and increase the cell migration ability, whereas the overexpression of MILI in B16BL6 could inhibit MAGEA expression and decrease the cell migration ability. Further investigations showed that MILI can methylate LINE1, which is crucial for MAGEA expression and melanoma cell migration. Our results provide a novel function of MILI in melanoma metastasis and tumor progression.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hidróxido de sodio, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hidróxido de sodio, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio solution, 50% in H2O
Sigma-Aldrich
Hidróxido de sodio solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Hidróxido de sodio solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hidróxido de sodio, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, powder
Sigma-Aldrich
Hidróxido de sodio, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Hidróxido de sodio solution, 5.0 M
Sigma-Aldrich
Hidróxido de sodio, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, flakes
Supelco
Hidróxido de sodio solution, 49-51% in water, eluent for IC
Supelco
Hidróxido de sodio solution, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hidróxido de sodio, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hidróxido de sodio, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Hydroquinone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydroquinone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hidróxido de sodio, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Supelco
Hydroquinone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O