Merck
The cancer pain related factors affected by celecoxib together with cetuximab in head and neck squamous cell carcinoma.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-03-18)
Yaqiong Yang, Jia Yan, Yan Huang, Hui Xu, Ying Zhang, Rong Hu, Jue Jiang, Zhifeng Chen, Hong Jiang
RESUMEN
Pain is the most disruptive influence on the quality of prognosis among head and neck squamous cell carcinoma (HNSCC) patients. The development of pain is closely associated with tumor growth and inflammation in the cancer patients. Notably, cyclooxygenase-2 (COX-2) is an important mediator during inflammation. Celecoxib, a selective inhibitor of COX-2, was hailed as a promising chemopreventive agent for HNSCC. Dose-dependent cardiac toxicity limits long-term use of celecoxib. However, the toxicity can be diminished by lowering the dosage. In this study, we hypothesized that a combinatory strategy to reduce cancer pain via two distinct pathways, tumor grown inhibition and inflammation blockade, which would enhance analgesia effect induced by HNSCC. We found that treatment of cetuximab (C225), a monoclonal anti-epidermal growth factor receptor (EGFR) antibody, with low-dose celecoxib results in a more pronounced anticancer effect in HNSCC than either agent alone. More noticeably, the combination could downregulate the phosphorylation of constitutively active extracellular signal regulated kinase (ERK) in CAL-27 and Fadu cells. Furthermore, combination therapy enhancing S phase arrest and downregulating cyclin D1 was observed in Fadu cells. The COX-2 expression was significantly blocked by celecoxib combined with C225, and other cancer pain related factors, such as ET-1 and NGF, was also downregulated by combination treatment. Taken together, these results strongly suggest that combination of celecoxib with C225 holds potential as a new therapy strategy in developing cancer pain treatment in HNSCC.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Dimetilsulfóxido, analytical standard
Supelco
Dimetilsulfóxido, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimetilsulfóxido, United States Pharmacopeia (USP) Reference Standard
Dimetilsulfóxido, European Pharmacopoeia (EP) Reference Standard
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimetilsulfóxido, ≥99.6%, ReagentPlus®