Merck
Extraction, characterization and in vitro antioxidant activity of polysaccharides from black soybean.
International journal of biological macromolecules (2014-09-27)
Jun Liu, Xiao-yuan Wen, Xue-qing Zhang, Hui-min Pu, Juan Kan, Chang-hai Jin
RESUMEN
Optimization of extraction conditions, preliminary characterization and in vitro antioxidant activity of polysaccharides from black soybean (BSPS) were investigated. The results of Box-Behnken design showed that the optimal extraction conditions for BSPS were as follows: ratio of water to material of 20 ml/g, extraction time of 6.4h and extraction temperature of 92 °C, with a corresponding yield of 2.56%. The crude BSPS were further fractionated on DEAE-52 and Sepharose CL-4B chromatography to afford three purified fractions (BSPS-1, BSPS-2 and BSPS-3). Chemical analysis showed that the three purified fractions were mainly composed of carbohydrate and uronic acid. In addition, BSPS-1 was composed of arabinose, rhamnose, galactose, glucose and mannose in the molar ratio of 1.79:1.00:2.59:26.54:1.01. BSPS-2 was composed of arabinose, rhamnose, xylose, galactose and mannose in the molar ratio of 8.10:4.80:9.15:13.38:1.00. BSPS-3 was composed of arabinose, rhamnose, galactose and mannose in the molar ratio of 16.80:3.60:33.66:1.00. The results of Fourier transform-infrared spectroscopy further confirmed the characteristic polysaccharide structures of the three purified fractions. Moreover, antioxidant assays showed crude BSPS and its purified fractions had potential superoxide anion and DPPH radical scavenging activities, and their antioxidant activity decreased in the order of crude BSPS > BSPS-3 > BSPS-2 > BSPS-1.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2,2-difenil-1-picrilhidrazil
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
Ácido L-ascórbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Ácido L-ascórbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Clorotrimetilsilano, ≥98.0% (GC)
Sigma-Aldrich
D-(+)-Galactosa, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ácido L-ascórbico, 99%
Sigma-Aldrich
Ácido L-ascórbico, reagent grade, crystalline
Sigma-Aldrich
L-(−)-Fucosa, ≥98% (GC)
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Clorotrimetilsilano, purified by redistillation, ≥99%
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactosa, ≥99% (HPLC)
Sigma-Aldrich
Ácido L-ascórbico, ACS reagent, ≥99%
Sigma-Aldrich
D-(+)-Manosa, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%
Sigma-Aldrich
Ácido L-ascórbico, meets USP testing specifications
Sigma-Aldrich
Ácido L-ascórbico, reagent grade
Sigma-Aldrich
D-(+)-Galactosa, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Manosa, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
D-(+)-Manosa, ≥99% (GC), wood
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)