Merck
PTD-Modified ATTEMPTS for Enhanced Toxin-based Cancer Therapy: An In Vivo Proof-of-Concept Study.
Pharmaceutical research (2015-02-24)
Meong Cheol Shin, Jian Zhang, Kyoung Ah Min, Huining He, Allan E David, Yongzhuo Huang, Victor C Yang
RESUMEN
To investigate the feasibility of applying PTD-modified ATTEMPTS (Antibody Targeted Triggered Electrically Modified Prodrug-Type Strategy) for enhanced toxin therapy for the treatment of cancer. A heparin-functionalized murine anti-CEA monoclonal antibody (mAb), T84.66-heparin (T84.66-Hep), was chemically synthesized and characterized for specific binding to CEA overexpressed cells. The T84.66-Hep was then applied to the PTD-modified ATTEMPTS approach and the crucial features of the drug delivery system (DDS), 'antibody targeting' and 'heparin/protamine-based prodrug', were evaluated in vitro to examine whether it could selective delivery a PTD-modified toxin, recombinant TAT-gelonin chimera (TAT-Gel), to CEA high expression cancer cells (LS174T). Furthermore, the feasibility of the drug delivery system (DDS) was assessed in vivo by biodistribution and efficacy studies using LS174T s.c. xenograft tumor bearing mice. T84.66-Hep displayed specific binding, but limited internalization (35% after 48 h incubation) to CEA high expression LS174T cells over low expression HCT116 cells. When mixed together with TAT-Gel, the T84.66-Hep formed a strong yet reversible complex. This complex formation provided an effective means of active tumor targeting of TAT-Gel, by 1) directing the TAT-Gel to CEA overexpressed tumor cells and 2) preventing nonspecific cell transduction to non-targeted normal cells. The cell transduction of TAT-Gel could, however, be efficiently reversed by addition of protamine. Feasibility of in vivo tumor targeting and "protamine-induced release" of TAT-Gel from the T84.66-Hep counterpart was confirmed by biodistribution and preliminary efficacy studies. This study successfully demonstrated in vitro and in vivo the applicability of PTD-modified ATTEMPTS for toxin-based cancer therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
1,2-Dicloroetano, anhydrous, 99.8%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Rhodamine B isothiocyanate, mixed isomers
Sigma-Aldrich
Rhodamine B isothiocyanate, BioReagent, mixed isomers, suitable for protein labeling
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Ethanesulfonic acid, 95%
SAFC
Isopropyl β-D-1-thiogalactopyranoside