Merck
  • New Trend for Acceleration Solid Phase Extraction Process Based on Using Magnetic Nano-adsorbents along with Surface Functionalization through Microwave Assisted Solvent-free Technique.
New Trend for Acceleration Solid Phase Extraction Process Based on Using Magnetic Nano-adsorbents along with Surface Functionalization through Microwave Assisted Solvent-free Technique.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-10-16)
Salwa A Ahmed, Ezzat M Soliman
RESUMEN
The use of a microwave assisted solvent-free technique for silica coating of iron magnetic nanoparticles (Fe3O4-MNPs) and their functionalization with three aliphatic diamines: 1,2-ethylenediamine (1,2EDA), 1,5-pentanediamine (1,5PDA) and 1.8-octanediamine (1,8-ODA), were successfully achieved in a very short time. Only 60 min were needed for the nano-adsorbent modification as compared with more than 1000 min using conventional methods under reflux conditions. Their surface characteristics (observed by TEM, XRD and FT-IR), in addition to Cu(II) adsorption capacities (1.805, 1.928 and 2.116 mmol g(-1)) and time of equilibration (5 s) were almost the same. Thus, the time required to accomplish the solid phase extraction process is greatly reduced. On the other hand, the phenomenon of the fast equilibration kinetics was successfully extended on using the functionalized aliphatic diamines magnetic nano-adsorbents as precursors for further microwave treatment. Three selective magnetic nano-adsorbents (Fe3O4-MNPs-SiO2-1,2EDA-3FSA, Fe3O4-MNPs-SiO2-1,5PDA-3FSA and Fe3O4-MNPs-SiO2-1,8ODA-3FSA) were obtained via the reaction with 3-formayl salicylic acid (3FSA) as a selective reagent for Fe(III). At 5 s contact time, they exhibited maximum Fe(III) uptake equal to 4.512, 4.987 and 5.367 mmol g(-1), respectively. Furthermore, modeling of values of metal uptake capacity obtained at different shaking time intervals supports pseudo-second order kinetics.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Hidróxido de amonio solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Hidróxido de sodio, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hidróxido de sodio, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio solution, 50% in H2O
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Cloruro de hidrógeno solution, 4.0 M in dioxane
Sigma-Aldrich
Hidróxido de amonio solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Cloruro de hierro (III) hexahydrate, ACS reagent, 97%
Sigma-Aldrich
Hidróxido de sodio solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Hidróxido de sodio solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Cloruro de hierro (III) hexahydrate, reagent grade, ≥98%, chunks
Sigma-Aldrich
Hidróxido de sodio, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Ácido clorhídrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ácido clorhídrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cloruro de hidrógeno solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, powder
Sigma-Aldrich
Cloruro de hierro (III) hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hidróxido de sodio, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Supelco
Ácido clorhídrico solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hidróxido de sodio solution, 5.0 M
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade
Sigma-Aldrich
Hidróxido de sodio, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, flakes
Sigma-Aldrich
Cloruro de hidrógeno solution, 1.0 M in diethyl ether