Merck
Activation of μ opioid receptors modulates inflammation in acute experimental colitis.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2015-02-19)
L Anselmi, J Huynh, C Duraffourd, I Jaramillo, G Vegezzi, F Saccani, E Boschetti, N C Brecha, R De Giorgio, C Sternini
RESUMEN
μ opioid receptors (μORs) are expressed by neurons and inflammatory cells, and mediate immune response. We tested whether activation of peripheral μORs ameliorates the acute and delayed phase of colitis. C57BL/6J mice were treated with 3% dextran sodium sulfate (DSS) in water, 5 days with or without the peripherally acting μOR agonist, [D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin (DAMGO) or with DAMGO+μOR antagonist at day 2-5, then euthanized. Other mice received DSS followed by water for 4 weeks, or DSS with DAMGO starting at day 2 of DSS for 2 or 3 weeks followed by water, then euthanized at 4 weeks. Disease activity index (DAI), histological damage, and myeloperoxidase assay (MPO), as index of neutrophil infiltration, were evaluated. Cytokines and μOR mRNAs were measured with RT-PCR, and nuclear factor-kB (NF-kB), the antiapoptotic factor Bcl-xL, and caspase 3 and 7 with Western blot. DSS induced acute colitis with elevated DAI, tissue damage, apoptosis and increased MPO, cytokines, μOR mRNA, and NF-kB. DAMGO significantly reduced DAI, inflammatory indexes, cytokines, caspases, and NF-kB, and upregulated Bcl-xL, effects prevented by μOR antagonist. In DSS mice plus 4 weeks of water, DAI, NF-kB, and μOR were normal, whereas MPO, histological damage, and cytokines were still elevated; DAMGO did not reduce inflammation, and did not upregulate Bcl-xL. μOR activation ameliorated the acute but not the delayed phase of DSS colitis by reducing cytokines, likely through activation of the antiapoptotic factor, Bcl-xL, and suppression of NF-kB, a potentiator of inflammation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Cloruro de hidrógeno solution, 4.0 M in dioxane
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, ≥98%
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, Molecular Biology, ≥99%
Sigma-Aldrich
Ácido clorhídrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ácido clorhídrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Cloruro de hidrógeno solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Ácido clorhídrico solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, BioXtra, ≥99%
Sigma-Aldrich
Cloruro de hidrógeno solution, 1.0 M in diethyl ether
Sigma-Aldrich
Ácido clorhídrico solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Cloruro de hidrógeno solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Bromuro de hexadeciltrimetilamonio, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Ácido clorhídrico solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
1-Aminobenzotriazole
Sigma-Aldrich
Cloruro de hidrógeno solution, 1.0 M in acetic acid
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Aminobenztropine, solid