- Tetra-nickel substituted polyoxotungsate as an efficient sorbent for the isolation of His6-tagged proteins from cell lysate.
Tetra-nickel substituted polyoxotungsate as an efficient sorbent for the isolation of His6-tagged proteins from cell lysate.
Talanta (2017-05-30)
Meng-Meng Wang, Qing Chen, Dan-Dan Zhang, Xu-Wei Chen, Ming-Li Chen
PMID28551125
RESUMEN
By virtue of the flexible structure of polyoxometalates, Ni
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Albúmina from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder