Merck
The effects of low level laser therapy on both HIV-1 infected and uninfected TZM-bl cells.
Journal of biophotonics (2017-01-28)
Masixole Yvonne Lugongolo, Sello Lebohang Manoto, Saturnin Ombinda-Lemboumba, Malik Maaza, Patience Mthunzi-Kufa
RESUMEN
Human immunodeficiency virus (HIV-1) infection remains a major health problem despite the use of highly active antiretroviral therapy (HAART), which has greatly reduced mortality rates. Due to the unavailability of an effective vaccine and treatment that would completely eradicate the virus in infected individuals, the quest for new therapies continues. Low level laser therapy (LLLT) involves the exposure of cells to low levels of red or infrared light. LLLT has been widely used in different medical conditions, but not in HIV-1 infection. This study aimed to determine the effects of LLLT on HIV-1 infected and uninfected TZM-bl cells. Both infected and uninfected cells were irradiated at a wavelength of 660 nm with different fluences from 2 J/cm
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Medio de Eagle modificado de Dulbecco, glucosa elevada, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Disolución de tripsina-EDTA, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
DEAE-Dextran hydrochloride, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, L-glutamine 200 mM, streptomycin 10 mg/mL, penicillin 10,000 units, 0.1 μm filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad