Skip to Content
Merck

Skip To

AABH97CD7B60

AA BLOCKS, INC.

rac N,O-Didesmethyl Venlafaxine

Synonym(s):

4-[1-(1-hydroxycyclohexyl)-2-(methylamino)ethyl]phenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Ships in 21 days - from Aldrich Partner.
€259.37
25 mg
Ships in 21 days - from Aldrich Partner.
€544.58

About This Item

Linear Formula:
C15H23O2N1
CAS Number:
Molecular Weight:
249.35
MDL number:

Vendor SKU: AA009E0C-5MG

€259.37


Ships in 21 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H23NO2/c1-16-11-14(12-5-7-13(17)8-6-12)15(18)9-3-2-4-10-15/h5-8,14,16-18H,2-4,9-11H2,1H3

Inchi Key

MMSWXJSQCAEDLK-UHFFFAOYSA-N

Purity

100%

Storage temp.

N/A

Country of Origin

US


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AABH30372286AABH303721B9AABH9A21A3F4
description

-

description

-

description

-

description

-

Purity

100%

Purity

-

Purity

98%

Purity

100%

Inchi Key

MMSWXJSQCAEDLK-UHFFFAOYSA-N

Inchi Key

CJNMZHKSKAPHPA-UHFFFAOYSA-N

Inchi Key

KYYIDSXMWOZKMP-HNNXBMFYSA-N

Inchi Key

KYYIDSXMWOZKMP-OAHLLOKOSA-N

Storage temp.

N/A

Storage temp.

-20C

Storage temp.

N/A

Storage temp.

N/A

Country of Origin

US

Country of Origin

US

Country of Origin

US

Country of Origin

US

Inchi Code

1S/C15H23NO2/c1-16-11-14(12-5-7-13(17)8-6-12)15(18)9-3-2-4-10-15/h5-8,14,16-18H,2-4,9-11H2,1H3

Inchi Code

1S/C14H21NO2.ClH/c15-10-13(11-4-6-12(16)7-5-11)14(17)8-2-1-3-9-14;/h4-7,13,16-17H,1-3,8-10,15H2;1H

Inchi Code

1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/t15-/m0/s1

Inchi Code

1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/t15-/m1/s1




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library